ASTM D3606 calibration standard #4 without internal standard (2 components)
Lieferant: Restek
30650
REST30650EA 216 EUR
REST30650
ASTM D3606 calibration standard #4 without internal standard (2 components)
Standards Mineralölindustrie-Standards
The mix consists of benzene (0,67% vol/vol), toluene (5% vol/vol).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.