ASTM D3606 calibration standard #6 without internal standard (2 components)
Lieferant: Restek
30652
REST30652EA 216 EUR
REST30652
ASTM D3606 calibration standard #6 without internal standard (2 components)
Standards Mineralölindustrie-Standards
Contains two components, those are benzene and toluene.
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.