ASTM D3606 calibration standard #4 with sec-butanol internal standard (3 components)
Lieferant: Restek
30664
REST30664EA 38 EUR
REST30664
ASTM D3606 calibration standard #4 with sec-butanol internal standard (3 components)
Standards Mineralölindustrie-Standards
Consists of three components, those are benzene, 2-butanol and toluene.
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.