ASTM® D3606 calibration standard #5 with sec-butanol internal standard (3 components)
Lieferant: Restek
30665
REST30665EA
38
EUR
REST30665
ASTM® D3606 calibration standard #5 with sec-butanol internal standard (3 components)
Standards
Mineralölindustrie-Standards
Consists of three components, those are benzene, 2-butanol and toluene.
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.