ASTM D3606 calibration standard #6 with sec-butanol internal standard (3 components)
Lieferant: Restek
30666
REST30666EA
38
EUR
REST30666
ASTM D3606 calibration standard #6 with sec-butanol internal standard (3 components)
Standards
Mineralölindustrie-Standards
Consists of three components, those are benzene, 2-butanol and toluene.
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.