SV calibration mix #2 (15 components)
Lieferant: Restek
31008
REST31008EA 51 EUR
REST31008
SV calibration mix #2 (15 components)
Standards Organische Standards
SV calibration mix #2 contains benzoic acid, 4-chloro-3-methylphenol, 2-chlorophenol, 2,4-dichlorophenol, 2,4-dimethylphenol, 4,6-dinitro-2-methylphenol (Dinitro-o-cresol), 2,4-dinitrophenol, 2-methylphenol (o-cresol), 4-methylphenol (p-cresol), 2-nitrophenol, 4-nitrophenol, pentachlorophenol, phenol, 2,4,5-trichlorophenol, 2,4,6-trichlorophenol.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.