SV calibration mix #5/610 PAH mix (16 components)
Lieferant: Restek
31011
REST31011EA
177.5
EUR
REST31011
SV calibration mix #5/610 PAH mix (16 components)
Standards
Organische Standards
SV calibration mix contains acenaphthene, acenaphthylene, anthracene, benz(a)anthracene, benzo(a)pyrene, benzo(b)fluoranthene, benzo(k)fluoranthene, benzo(ghi)perylene, chrysene, dibenz(a,h)anthracene, fluoranthene, fluorene, indeno(1,2,3-cd)pyrene, naphthalene, phenanthrene, pyrene.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.