SV calibration mix #6 (18 components)
Lieferant: Restek
31012
REST31012EA
255.5
EUR
REST31012
SV calibration mix #6 (18 components)
Standards
Organische Standards
sv calibration mix #6 contains aldrin, α-bhc, β-bhc, δ-bhc, γ-bhc (lindane), 4,4'-DDD, 4,4'-DDE, 4,4'-DDT, dieldrin, endosulphan I, endosulphan II, endosulphan sulphate, endrin, endrin aldehyde, endrin ketone, heptachlor, heptachlor epoxide (isomer B), methoxychlor.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.