TCLP Acid mix (6 components)
Lieferant: Restek
31027
REST31027EA
44.5
EUR
REST31027
TCLP Acid mix (6 components)
Standards
Organische Standards
TCLP acid mix contains 2-methylphenol (o-cresol), 3-methylphenol (m-cresol), 4-methylphenol (p-cresol), pentachlorophenol, 2,4,5-trichlorophenol, 2,4,6-trichlorophenol.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.