611 Haloethers calibration mix (5 components)
Lieferant: Restek
31034
REST31034EA
68
EUR
REST31034
611 Haloethers calibration mix (5 components)
Standards
Umweltanalyse-Standards
Haloethers calibration mix contains bis(2-chloroethoxy)methane, bis(2-chloroethyl)ether, 4-bromophenyl phenyl ether, 4-chlorophenyl phenyl ether, 2,2'-oxybis(1-chloropropane).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.