612 Chlorinated hydrocarbons calibration mix (9 components)
Lieferant: Restek
31035
REST31035EA
57.5
EUR
REST31035
612 Chlorinated hydrocarbons calibration mix (9 components)
Standards
Umweltanalyse-Standards
612 Chlorinated hydrocarbons calibration mix contains 2-chloronaphthalene, 1,2-dichlorobenzene, 1,3-dichlorobenzene, 1,4-dichlorobenzene, hexachlorobenzene, hexachlorobutadiene, hexachlorocyclopentadiene, hexachloroethane, 1,2,4-trichlorobenzene.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.