625 kit
Lieferant: Restek
625 kit contains 604 phenols calibration mix: 4-chloro-3-methylphenol (59-50-7), 2-chlorophenol (95-57-8), 2,4-dichlorophenol (120-83-2), 2,4-dimethylphenol (105-67-9), 4,6-dinitro-2-methylphenol (dinitro-o-cresol) (534-52-1), 2,4-dinitrophenol (51-28-5), 2-nitrophenol (88-75-5), 4-nitrophenol (100-02-7), pentachlorophenol (87-86-5), phenol (108-95-2), 2,4,6-trichlorophenol (88-06-2), benzidines calibration mix: benzidine (92-87-5), 3,3'-dichlorobenzidine (91-94-1), 606 phthalate esters calibration mix: bis(2-ethylhexyl)phthalate (117-81-7), benzyl butyl phthalate (85-68-7), di-n-butyl phthalate (84-74-2), diethylphthalate (84-66-2), dimethylphthalate (131-11-3), di-n-octyl phthalate (117-84-0),31032: 607 nitrosamines calibration mix, n-nitrosodimethylamine (62-75-9), n-nitroso-di-n-propylamine (621-64-7), n-nitrosodiphenylamine (86-30-6),31033: 609 nitroaromatics & isophorone calibration mix: 2,4-dinitrotoluene (121-14-2), isophorone (78-59-1), 2,6-dinitrotoluene (606-20-2), nitrobenzene (98-95-3),31011: sv calibration mix #5/610 pah mix: acenaphthene (83-32-9), acenaphthylene (208-96-8), anthracene (120-12-7), benz(a)anthracene (56-55-3), benzo(a)pyrene (50-32-8), benzo(b)fluoranthene (205-99-2), benzo(k)fluoranthene (207-08-9), benzo(ghi)perylene (191-24-2), chrysene (218-01-9), dibenz(a,h)anthracene (53-70-3), fluoranthene (206-44-0), fluorene (86-73-7), indeno(1,2,3-cd)pyrene (193-39-5), naphthalene (91-20-3), phenanthrene (85-01-8), pyrene (129-00-0),31034: 611 haloethers calibration mix, bis(2-chloroethoxy)methane (111-91-1), bis(2-chloroethyl)ether (111-44-4), 4-bromophenyl phenyl ether (101-55-3), 4-chlorophenyl phenyl ether (7005-72-3), 2,2'-oxybis(1-chloropropane) (108-60-1),31035: 612 chlorinated hydrocarbons calibration mix: 2-chloronaphthalene (91-58-7), 1,2-dichlorobenzene (95-50-1), 1,3-dichlorobenzene (541-73-1), 1,4-dichlorobenzene (106-46-7), hexachlorobenzene (118-74-1), hexachlorobutadiene (87-68-3), hexachlorocyclopentadiene (77-47-4), hexachloroethane (67-72-1), 1,2,4-trichlorobenzene (120-82-1).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Bestellinformation: Because most laboratories do not routinely analyse pesticides, PCBs, toxaphene, and chlordane in their calibration mixtures for Method 625, these mixtures are not included in the 625 kit. They may be purchased separately or in the 608 complete kit.