Pharmaceuticals Mix #1 (8 components)
Lieferant: Restek
31116
REST31116EA
80.5
EUR
REST31116
Pharmaceuticals Mix #1 (8 components)
Standards
Organische Standards
This pharmaceuticals mix #1 contain eight components: Acetaminophen, Caffeine, Carbamazepine, Ciprofloxacin HCL, Erythromycin USP, Fluoxetine HCl, Sulphamethoxazole and Trimethoprim.
Shelf life 18 months.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.