Fuel Oil #4 Standards (ASTM D-445)
Lieferant: Restek
Fuel oil #4 is typically used in limited applications in which the fuel cannot be preheated prior to burning. The fuel is a blend of distillate (fuel oil #2) and residual (fuel oil #6).
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.