Calibration mix D2887 (17 components)
Lieferant: Restek
D2887 calibration mix contains compound, conc.; (C6) n-hexane (110-54-3), 6% w/w; (C7) n-heptane (142-82-5), 6% w/w; (C8) n-octane (111-65-9), 8% w/w; (C9) n-nonane (111-84-2), 8% w/w; (C10) n-decane (124-18-5), 12% w/w; (C11) n-undecane (1120-21-4), 12% w/w; (C12) n-dodecane (112-40-3), 12% w/w; (C14) n-tetradecane (629-59-4), 12% w/w; (C16) n-hexadecane (544-76-3), 10% w/w; (C18) n-octadecane (593-45-3), 5% w/w; (C20) n-eicosane (112-95-8), 2% w/w; (c24) n-tetracosane (646-31-1), 2% w/w; (c28) n-octacosane (630-02-4), 1% w/w; (C32) n-dotriacontane (544-85-4), 1% w/w; (C36) n-hexatriacontane (630-06-8), 1% w/w; (C40) n-tetracontane (4181-95-7), 1% w/w; (C44) n-tetratetracontane (7098-22-8), 1% w/w.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.