Weathered diesel fuel #2 kit
Lieferant: Restek
31239
REST31239EA
210.5
EUR
REST31239
Weathered diesel fuel #2 kit
Standards
Mineralölindustrie-Standards
The kit consists of different standards.
- Contains 1 ml each of these mixtures,
- Diesel fuel #2 standard
- Diesel fuel #2 standard - 25% weathered
- Diesel fuel #2 standard - 50% weathered
- Diesel fuel #2 standard - 75% weathered
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...