610 PAH calibration mix A (16 components)
Lieferant: Restek
610 PAH calibration mix A contains acenaphthene (1000 μg/ml), acenaphthylene (1000 μg/ml),anthracene (1000 μg/ml), benz(a)anthracene (500 μg/ml), benzo(a)pyrene (500 μg/ml), benzo(b)fluoranthene (500 μg/ml)
benzo(k)fluoranthene (500 μg/ml), benzo(ghi)perylene (500 μg/ml), chrysene (500 μg/ml), dibenz(a,h)anthracene (500 μg/ml), fluoranthene (500 μg/ml), fluorene (1000 μg/ml), indeno(1,2,3-cd)pyrene (500 μg/ml), naphthalene (1000 μg/ml), phenanthrene (500 μg/ml), pyrene (500 μg/ml).
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.