Method 1664 oil and grease mix (2 components)
Lieferant: Restek
31457
REST31457EA 47.5 EUR
REST31457
Method 1664 oil and grease mix (2 components)
Standards Mineralölindustrie-Standards
Contains two components, those are n-hexadecane and stearic acid.
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.