MA EPH aliphatic hydrocarbon standard (14 components)
Lieferant: Restek
31459
REST31459EA 79.5 EUR
REST31459
MA EPH aliphatic hydrocarbon standard (14 components)
Standards Umweltanalyse-Standards
Contains: n-nonane, C9, n-decane, C10, n-dodecane, C12, n-tetradecane, C14, n-hexadecane, C16, n-octadecane, C18, n-nonadecane, C19, n-eicosane, C20, n-docosane, C22, n-tetracosane, C24, n-hexacosane, C26, n-octacosane, C28, n-triacontane, C30, n-hexatriacontane, C36.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.