TX TPH matrix spike mix (2 components)
Lieferant: Restek
31484
REST31484EA 52 EUR
REST31484
TX TPH matrix spike mix (2 components)
Standards Mineralölindustrie-Standards
The mix consists of diesel fuel #2 composite, unleaded gasoline composite.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.