WA EPH aromatic hydrocarbon mix (6 components)
Lieferant: Restek
31488
REST31488EA
72.5
EUR
REST31488
WA EPH aromatic hydrocarbon mix (6 components)
Standards
Mineralölindustrie-Standards
The mix consists of, (C8) n-octane, (C10) n-decane, (C12) n-dodecane, (C16) n-hexadecane, (C21) n-heneicosane, (C34) n-tetratriacontane.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.