WA EPH aliphatic hydrocarbon mix (6 components)
Lieferant: Restek
31489
REST31489EA 52 EUR
REST31489
WA EPH aliphatic hydrocarbon mix (6 components)
Standards Mineralölindustrie-Standards
The mix consists of, (C8) n-octane, (C10) n-decane, (C12) n-dodecane, (C16) n-hexadecane, (C21) n-heneicosane, (C34) n-tetratriacontane.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.