CLP 04.1 B/N matrix spike mix (4 components)
Lieferant: Restek
31492
REST31492EA 64 EUR
REST31492
CLP 04.1 B/N matrix spike mix (4 components)
Standards Organische Standards
The mix consists of acenaphthene, 2,4-dinitrotoluene, N-nitroso-di-n-propylamine, pyrene.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.