CLP 04.1 phenols calibration mix (14 components)
Lieferant: Restek
31494
REST31494EA
97
EUR
REST31494
CLP 04.1 phenols calibration mix (14 components)
Standards
Organische Standards
The mix consists of 4-chloro-3-methylphenol, 2-chlorophenol, 2,4-dichlorophenol, 2,4-dimethylphenol, 4,6-dinitro-2-methylphenol, 2,4-dinitrophenol, 2-methylphenol, 4-methylphenol, 2-nitrophenol, 4-nitrophenol, pentachlorophenol, phenol, 2,4,5-trichlorophenol, 2,4,6-trichlorophenol.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.