1,2-Diphenylhydrazine
Lieferant: Restek
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.
Bestellinformation: Note that 1,2-diphenylhydrazine is an unstable compound that will oxidize to azobenzene, thereby decreasing the concentration of 1,2-diphenylhydrazine over time. For accurate calibration results, it is recommended that the concentrations of 1,2-diphenylhydrazine and azobenzene be combined. Please contact Restek Technical Service if you have any questions about this issue.