Single-component explosives solutions
Lieferant: Restek
31452
31453
31498
31600
31601
31602
31657
31658
31659
31660
31661
31662
31663
31664
31665
31666
31668
31669
31670
31671
33901
REST31452EA
52
EUR
REST31452
REST31453
REST31498
REST31600
REST31601
REST31602
REST31657
REST31658
REST31659
REST31660
REST31661
REST31662
REST31663
REST31664
REST31665
REST31666
REST31668
REST31669
REST31670
REST31671
REST33901
Single-component explosives solutions
Standards
Organische Standards
Single-component explosives solutions supports US department of defense base closures and remediation.
- Mixtures and singles supporting LC US EPA method 8330
- Mixtures and singles supporting GC-ECD US EPA method 8095
- Internal standards and surrogates to support both methods
These materials support nitroaromatic, nitramine, and nitroester analyses by GC-ECD (Method 8095). Compounds listed are explosives, manufacturing intermediates, or degradation products. Method 8095 mixtures contains the components at concentration ratios appropriate for ECD.
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)