8095 calibration mix A (10 components)
Lieferant: Restek
31607
REST31607EA
93
EUR
REST31607
8095 calibration mix A (10 components)
Standards
Organische Standards
The mix consists of, 2-amino-4,6-dinitrotoluene, 4-amino-2,6-dinitrotoluene, 1,3-dinitrobenzene, 2,4-dinitrotoluene, 2,6-dinitrotoluene, HMX, RDX, tetryl, 1,3,5-trinitrobenzene, 2,4,6-trinitrotoluene.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.