8095 calibration mix B (7 components)
Lieferant: Restek
31608
REST31608EA
80
EUR
REST31608
8095 calibration mix B (7 components)
Standards
Organische Standards
The mix consists of, 3,5-dinitroaniline, nitrobenzene, nitroglycerin, 2-nitrotoluene, 3-nitrotoluene, 4-nitrotoluene, PETN.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.