8095 Matrix spike mix B (7 components)
Lieferant: Restek
31610
REST31610EA 49 EUR
REST31610
8095 Matrix spike mix B (7 components)
Standards Organische Standards
The mix consists of, 3,5-dinitroaniline, nitrobenzene, nitroglycerin, 2-nitrotoluene, 3-nitrotoluene, 4-nitrotoluene, PETN.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.