Calibration mixture, Connecticut ETPH
Lieferant: Restek
31614
REST31614EA 79.5 EUR
REST31614
Calibration mixture, Connecticut ETPH
Standards Gaschromatographie-Standards
Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs.
  • 15 components- all n- alkanes