Certified PAHs in diesel
Lieferant: Restek
31673
REST31673EA 0 EUR
REST31673
Certified PAHs in diesel
Standards Mineralölindustrie-Standards
The mix consists of, acenaphthene, 2-methylnaphthalene, acenaphthylene, fluorene, naphthalene, 1-methylnaphthalene and phenanthrene.
  • Confirms diesel #2 TPH and priority PAHs in a single analysis
  • Complete data packs available
Certificate of analysis includes concentration of TPH and certified concentrations of individual PAHs.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.