Extraction solvent stock solution #2 (2 components)
Lieferant: Restek
31680
REST31680EA 114.5 EUR
REST31680
Extraction solvent stock solution #2 (2 components)
Standards Umweltanalyse-Standards
This extraction solvent stock solution contains: n-decane (C10) (20 µl/l), n-tetracontane (C40) (20 mg/l).