SV system performance check mix (4 components)
Lieferant: Restek
31689
REST31689EA
69.5
EUR
REST31689
SV system performance check mix (4 components)
Standards
Organische Standards
This SV system performance check mix consist of four components: 2,4-Dinitrophenol, Hexachlorocyclopentadiene, 4-Nitrophenol and N-Nitroso-di-n-propylamine.
Shelf life 6 to 24 months.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.