Phenol calibration mix, EPA 528
Lieferant: Restek
31694
REST31694EA
71
EUR
REST31694
Phenol calibration mix, EPA 528
Standards
Organische Standards
This phenol calibration mix contain 12 components: 4-Chloro-3-methylphenol, 2-Chlorophenol, 2,4-Dichlorophenol, 2,4-Dimethylphenol, 4,6-Dinitro-2-methylphenol (Dinitro-o-cresol), 2,4-Dinitrophenol, 2-Methylphenol (o-cresol), 2-Nitrophenol, 4-Nitrophenol, Pentachlorophenol, Phenol and 2,4,6-Trichlorophenol.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.