Aldehyde-ketone-DNPH TO-11A calibration mix (15 components)
Lieferant: Restek
31808
REST31808EA
126
EUR
REST31808
Aldehyde-ketone-DNPH TO-11A calibration mix (15 components)
Standards
Organische Standards
This consists of acetaldehyde-DNPH; acetone-DNPH; acrolein-DNPH; benzaldehyde-DNPH; n-butyraldehyde-DNPH; crotonaldehyde-DNPH; 2,5-dimethylbenzaldehyde-DNPH; formaldehyde-DNPH; hexaldehyde-DNPH; isovaleraldehyde-DNPH; propionaldehyde-DNPH; m-tolualdehyde-DNPH; o-Tolualdehyde-DNPH; p-tolualdehyde-DNPH and aleraldehyde-DNPH.
Bitte beachten *The reported concentrations reflect the amount of aldehyde or ketone in the mixture. The concentration of derivatized aldehyde or ketone is not reported.