TNRCC 1006 retention time marker mix (9 components)
Lieferant: Restek
31814
REST31814EA 72.5 EUR
REST31814
TNRCC 1006 retention time marker mix (9 components)
Standards Umweltanalyse-Standards
This TNRCC 1006 retention time marker mix contains: (C6) n-Hexane, (C7) n-Heptane, (C8) n-Octane, (C10) n-Decane, (C12) n-Dodecane, (C16) n-Hexadecane, (C21) n-Heneicosane, (C28) n-Octacosane, (C35) n-Pentatriacontane.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.