Residual range calibration standard (RCS)
Lieferant: Restek
31817
REST31817EA 51 EUR
REST31817
Residual range calibration standard (RCS)
Standards Mineralölindustrie-Standards
SAE30 motor oil:SAE40 motor oil (1:1).
  • 2 components