Method 525.2 internal standard mix
Lieferant: Restek
31825
REST31825EA 62 EUR
REST31825
Method 525.2 internal standard mix
Standards Umweltanalyse-Standards
This mix contains 3 components: acenaphthene-d10, chrysene-d12 and phenanthrene-d10.
Standards: Certified reference materials (CRMs) manufactured and QC tested in ISO-accredited labs satisfy your ISO requirements.