Method 525.2 surrogate standard mix
Lieferant: Restek
31826
REST31826EA 46.5 EUR
REST31826
Method 525.2 surrogate standard mix
Standards Umweltanalyse-Standards
This mix contains 2-nitro-m-xylene, perylene-d12, pyrene-d10 and triphenylphosphate.