DRO/ORO range calibration standard, (12 components)
Lieferant: Restek
31832
REST31832EA
74.5
EUR
REST31832
DRO/ORO range calibration standard, (12 components)
Standards
Mineralölindustrie-Standards
This consists of n-decane (C10), n-dodecane (C12), n-tetradecane (C14), n-hexadecane (C16), n-octadecane (C18), n-eicosane (C20) ,n-docosane (C22) , n-tetracosane (C24), n-hexacosane (C26), n-octacosane (C28), n-triacontane (C30) and n-dotriacontane (C32).