Creosote oil standard
Lieferant: Restek
31838
REST31838EA
79.5
EUR
REST31838
Creosote oil standard
Standards
Mineralölindustrie-Standards
This is for total petroleum hydrocarbon pattern recognition of creosote oil.
Creosote oil, a widely used wood preservative produced by distilling coal tar, contains chemicals that are classified as carcinogens (benzo(a)pyrene).