506 Calibration mix (4 components)
Lieferant: Restek
31845
REST31845EA 60 EUR
REST31845
506 Calibration mix (4 components)
Standards Organische Standards
The mix consists of benzyl butyl phthalate, bis(2-ethylhexyl)adipate, bis(2-ethylhexyl)phthalate, di-n-butylphthalate, diethylphthalate, dimethylphthalate, di-n-octyl phthalate.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.