PAH supplement mix for method 8100 (8 components)
Lieferant: Restek
31857
REST31857EA 569 EUR
REST31857
PAH supplement mix for method 8100 (8 components)
Standards Organische Standards
The mix consists of benzo(j)fluoranthene, dibenz(a,h)acridine, dibenz(a,j)acridine, 7H-dibenzo(c,g)carbazole, dibenzo(a,e)pyrene, dibenzo(a,h)pyrene, dibenzo(a,i)pyrene, 3-methylcholanthrene.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.