n-Tetracontan

Lieferant: Restek

31859
REST31859EA 48.5 EUR
REST31859
n-Tetracontan
n-Tetracontane
Formel: H₃C(CH₂)₃₈CH₃
Molecular Weight: 563,09 g/mol
Schmelzpunkt: 82…83 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015267
CAS-Nummer: 4181-95-7
EINECS: 224-055-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR