Canadian drinking water triazine herbicides mix (7 components)
Lieferant: Restek
31864
REST31864EA
105.5
EUR
REST31864
Canadian drinking water triazine herbicides mix (7 components)
Standards
Umweltanalyse-Standards
The mix consists of alachlor, atrazine, cyanazine (bladex), metolachlor, metribuzin, prometryne, simazine.
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.