Canadian drinking water OP pesticides mix (9 components)
Lieferant: Restek
31867
REST31867EA
98
EUR
REST31867
Canadian drinking water OP pesticides mix (9 components)
Standards
Umweltanalyse-Standards
Consists of nine components, those are azinphos methyl, chlorpyrifos, diazinon, dimethoate, malathion, parathion, phorate, temephos and terbufos .
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.