Canadian drinking water phenoxyacid herbicides mix (11 components)
Lieferant: Restek
31868
REST31868EA 98 EUR
REST31868
Canadian drinking water phenoxyacid herbicides mix (11 components)
Standards Umweltanalyse-Standards
Consists of 11 components, those are bromoxynil, 2,4-d, dicamba, 2,4-dichlorophenol, diclofop methyl, dinoseb, pentachlorophenol, picloram, 2,4,5-t, 2,3,4,6-tetrachlorophenol and 2,4,6-trichlorophenol.
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.