EPA method 8310 PAH mixture
Lieferant: Restek
31874
REST31874EA
139.5
EUR
REST31874
EPA method 8310 PAH mixture
Standards
Umweltanalyse-Standards
EPA method 8310 PAH mixturecontain 18 components.
Matrix: Acetonitrile:toluene (92:8)
Standards: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.