SOM01.1 SVOA MegaMix standard, revised (65 components)
Lieferant: Restek
31900
REST31900EA 200.5 EUR
REST31900
SOM01.1 SVOA MegaMix standard, revised (65 components)
Standards Organische Standards
Consists of 65 components, those are acenaphthene, acenaphthylene, acetophenone, anthracene, benzanthracene, benzopyrene, benzofluoranthene, benzoperylene, benzofluoranthene, benzyl butyl phthalate, biphenyl, bismethane, bisether, bisphthalate, 4-bromophenyl-phenylether, carbazole, 4-chloroaniline, 4-chloro-3-methylphenol, 2-chloronaphthalene, 2-chlorophenol, 4-chlorophenyl-phenylether, chrysene, dibenzanthracene, dibenzofuran, 3,3'-dichlorobenzidine, 2,4-dichlorophenol, diethylphthalate, 2,4-dimethylphenol, dimethylphthalate, di-n-butylphthalate, 4,6-dinitro-2-methylphenol , 2,4-dinitrophenol, 2,4-dinitrotoluene, 2,6-dinitrotoluene, di-n-octyl phthalate, diphenylamine *,fluoranthene, fluorene, hexachlorobenzene, hexachloro-1,3-butadiene , hexachlorocyclopentadiene, hexachloroethane, indenopyrene, isophorone, 2-methylnaphthalene, 2-methylphenol , 3-methylphenol, 4-methylphenol , naphthalene, 2-nitroaniline, 3-nitroaniline, 4-nitroaniline, nitrobenzene, 2-nitrophenol, 4-nitrophenol, n-nitroso-di-n-propylamine, 2,2'-oxybis, pentachlorophenol, phenanthrene, phenol, pyrene, 1,2,4,5-tetrachlorobenzene, 2,3,4,6-tetrachlorophenol, 2,4,5-trichlorophenol and 2,4,6-trichlorophenol.
Standards: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Bestellinformation: *N-nitrosodiphenylamine (CLP-listed analyte) decomposes to diphenylamine (mix component) in the injector.