Reference standards for food analysis, Labeled pesticide residues
Lieferant: Restek
31984
31985
31987
31988
31989
31990
31986
REST31984EA
63
EUR
REST31984
REST31985
REST31987
REST31988
REST31989
REST31990
REST31986
Reference standards for food analysis, Labeled pesticide residues
Standards
Lebensmittel-Standards
These standards are used for pesticide residue quantification.
- Compatible with both LC-MS andGC-MS applications and for optimising LC-MS/MS system performance
- Would mitigate matrix effects
Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.