Reference standards for food analysis, Labeled pesticide residues
Lieferant: Restek
31984 31985 31987 31988 31989 31990 31986
REST31984EA 63 EUR
REST31984 REST31985 REST31987 REST31988 REST31989 REST31990 REST31986
Reference standards for food analysis, Labeled pesticide residues
Standards Lebensmittel-Standards
These standards are used for pesticide residue quantification.
  • Compatible with both LC-MS andGC-MS applications and for optimising LC-MS/MS system performance
  • Would mitigate matrix effects
Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.